Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NR2C2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-E46
RL100-401-E46EA 518 USD InStock
RL100-401-E46
Anti-NR2C2 Rabbit Polyclonal Antibody
Antibodies
Anti-TR4 (Rabbit) is suitable for use in Western Blotting. Anti-TR4 antibodies are specific for the ~64 kDa TR4 Protein by Western blot in cerebellum and nuclear extracts from MEL cell lines. Reactivity is expected from human and rat.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NR2C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty