Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-E81S
RL100-401-E81SEA 149 USD InStock
RL100-401-E81S
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Histone H3 antibody has been tested for use in ELISA and western blot. For western blots expect a band of approximately 15.4 kDa in size corresponding tothe Histone 3 Protein. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000;

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Xenopus laevis, Arabidopsis Thaliana, Caenorhabditis Elegans, Drosophila

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty