Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AHSA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-E82
RL100-401-E82EA 491 USD InStock
RL100-401-E82
Anti-AHSA1 Rabbit Polyclonal Antibody
Antibodies
Anti-Aha1 Antibody is suitable for use in WB, ELISA, IHC, Immunofluorescence microscopy and IP. Expect a band approximately ~38kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 0.5-1:1000;

Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty