Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-F49
RL100-401-F49EA 474 USD InStock
RL100-401-F49
Anti-HSPA8 Rabbit Polyclonal Antibody
Antibodies
Anti-Hsc70 (Hsp73) Antibody is suitable for use in WB, IF, IP, Immunochemistryand blocking. Expect a band approximately ~73kDa Protein corresponding to the molecular mass of hsc70 on SDS PAGE immunoblots.

Recommended Dilutions: Western Blot: 1: 1500-2000;

Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Hamster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty