Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPB8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-F54
RL100-401-F54EA 491 USD InStock
RL100-401-F54
Anti-HSPB8 Rabbit Polyclonal Antibody
Antibodies
Anti-Hsp22 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~22kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:1000;

Type: Primary
Antigen: HSPB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty