Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK14 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-G26
RL100-401-G26EA 522 USD InStock
RL100-401-G26
Anti-MAPK14 Rabbit Polyclonal Antibody
Antibodies
Anti-p38 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~43kDa Protein corresponding to the molecular mass of p38 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: User Optimizied; Immunoprecipitation: 1:250

Type: Primary
Antigen: MAPK14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat, Bovine, Rabbit, Pig, Dog, Hamster, Chicken, Sheep, Guinea Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty