Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDIA2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-G29
RL100-401-G29EA 522 USD InStock
RL100-401-G29
Anti-PDIA2 Rabbit Polyclonal Antibody
Antibodies
Anti-PDI Antibody is suitable for use in WB, IP, Immunofluorescence microscopy and IHC. Expect a band approximately ~58kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:4000;

Type: Primary
Antigen: PDIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Hamster, Monkey, Guinea Pig, Bovine, Sheep, Pig, Xenopus, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty