Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MORC3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-N96 100-401-N96
RL100-401-N96EA 518 USD InStock
RL100-401-N96 CA100-401-N96
Anti-MORC3 Rabbit Polyclonal Antibody
Antibodies
Anti-Morc3 Antibody is suitable for use in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 110.8 kDa in size corresponding Morc3 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: MORC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty