Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4147 100-4147
RL100-4147EA 1290 USD InStock
RL100-4147 CA100-4147
Anti-BAM1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-BETA AMYLASE (Sweet Potato) Reacts with Pan

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: BAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sweet Potato

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty