Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4165 100-4165
RL100-4165EA 518 USD InStock
RL100-4165 CA100-4165
Anti-RELA Rabbit Polyclonal Antibody
Antibodies
Anti-NFkB p65 (Rel A) is suitable for the detection by immunoblot of human and mouse NFkB p65 (Rel A), immunochemistry, and ELISA.  This product was also tested in a gel supershift assay and found to be reactive against all p65 (Rel A).

Recommended Dilutions: ELISA: 1:5000; GelShift: 05 µl - 1.0 µl; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:400;

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty