Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFKB cRel Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4166 100-4166
RL100-4166EA 518 USD InStock
RL100-4166 CA100-4166
Anti-NFKB cRel Rabbit Polyclonal Antibody
Antibodies
This product was assayed by immunoblot and found to be reactive against NFkB cRel at a dilution of 1:1000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; GelShift: 1:500; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: NFKB cRel
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty