Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFKBIA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4167C 100-4167C
RL100-4167CEA 518 USD InStock
CA100-4167C RL100-4167C
Anti-NFKBIA Rabbit Polyclonal Antibody
Antibodies
Anti-IkBa Antibody is suitable for western blotting, ELISA, and IHC. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty