Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ansB Rabbit Monoclonal Antibody [clone: 7C5G1H6H10]

Supplier: Rockland Immunochemicals
100-4171 100-4171
RL100-4171EA 1290 USD InStock
CA100-4171 RL100-4171
Anti-ansB Rabbit Monoclonal Antibody [clone: 7C5G1H6H10]
Antibodies
Primary Rabbit Anti-L-ASPARAGINASE (E.coli) Reacts with Pan

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ansB
Clonality: Monoclonal
Clone: 7C5G1H6H10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty