Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRSS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4180 100-4180
RL100-4180EA 1290 USD InStock
CA100-4180 RL100-4180
Anti-PRSS2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-TRYPSINOGEN (Bovine Pancreas) Reacts with Pan

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PRSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty