Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Dog serum Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
104-4101 104-4101
RL104-4101EA 313 USD InStock
CA104-4101 RL104-4101
Anti-Dog serum Rabbit Polyclonal Antibody
Antibodies
Anti-Canine serum is suitable for the specific deteciton of protiens found in dog serum. Cross reactivity to serum-protiens from other species has not been tested. Optimum experimental usage should be determined by researcher.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Dog serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty