Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Goat serum Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
105-4101 105-4101
RL105-4101EA 319 USD InStock
RL105-4101 CA105-4101
Anti-Goat serum Rabbit Polyclonal Antibody
Antibodies
Anti-goat serum antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Goat serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty