Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Golden Syrian Hamster serum Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
107-4101 107-4101
RL107-4101EA 319 USD InStock
RL107-4101 CA107-4101
Anti-Golden Syrian Hamster serum Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-GOLDEN SYRIAN HAMSTER SERUM Reacts with Hamster

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Golden Syrian Hamster serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Golden Syrian Hamster, Hamster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty