Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R2 Rabbit Monoclonal Antibody [clone: 8G1]

Supplier: Rockland Immunochemicals
109-401-305 109-401-305
RL109-401-305EA 1290 USD InStock
RL109-401-305 CA109-401-305
Anti-IL1R2 Rabbit Monoclonal Antibody [clone: 8G1]
Antibodies
This antiserum against Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA, radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.  IL-1RII has been termed p68 or CD121 in some laboratories.

Recommended Dilutions: ELISA: 1:1000 - 1:2000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:400 - 1:800;

Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 8G1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty