Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-312S
RL109-401-312SEA 148 USD InStock
RL109-401-312S
Anti-IL10 Rabbit Polyclonal Antibody
Antibodies
This antiserum has been tested for use in neutralizations, ELISA, immunochemistry, radioimmunoassay, immunoprecipitation, immunochemistry and immunoblotting. The antibody recognizes hrIL-10. Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: IL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty