Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNAJB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-E57
RL109-401-E57EA 491 USD InStock
RL109-401-E57
Anti-DNAJB1 Rabbit Polyclonal Antibody
Antibodies
Anti-Hsp40 Antibody is suitable for use in WB and IP. Expect a band approximately ~40kDa Protein corresponding to the molecular mass of Hsp40 on SDS PAGE immunoblots. Also detects recombinant Hsp40.

Recommended Dilutions: Western Blot: 1: 1500-2000

Type: Primary
Antigen: DNAJB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty