Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Mouse Serum Rabbit Polyclonal Antibody [clone: 2D11.H5]

Supplier: Rockland Immunochemicals
110-4101 110-4101
RL110-4101EA 319 USD InStock
CA110-4101 RL110-4101
Anti-Mouse Serum Rabbit Polyclonal Antibody [clone: 2D11.H5]
Antibodies
Anti-MOUSE SERUM (RABBIT) Antibody is suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Mouse Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty