Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GABRG2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
112-401-C81
RL112-401-C81EA 518 USD InStock
RL112-401-C81
Anti-GABRG2 Rabbit Polyclonal Antibody
Antibodies
Anti-GABA(A) Receptor gamma 2 (Rabbit) antibody is suitable for use in Western Blotting and IHC. Expect a band of approximately 46kDa in size corresponding 2-subunit of the GABAA receptor in Western blots of Rat brain extracts.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:400

Type: Primary
Antigen: GABRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty