Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PEBP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
112-401-E08
RL112-401-E08EA 518 USD InStock
RL112-401-E08
Anti-PEBP1 Rabbit Polyclonal Antibody
Antibodies
Anti-Phosphatidylethanolamie binding Protein Antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 22 kDa in size corresponding to the PEBP Protein.

Recommended Dilutions: Western Blot: 1:1000;

Type: Primary
Antigen: PEBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty