Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody [clone: eB182] (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
18-0216-32 18-0216-32
RL18-0216-32EA 292 USD InStock
RL18-0216-32 CA18-0216-32
Anti-IgG Mouse Monoclonal Antibody [clone: eB182] (FITC (Fluorescein Isothiocyanate))
Antibodies
Fluorescent Rabbit TrueBlot® Antibody Fluorescein is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB182
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty