Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rat Monoclonal Antibody [clone: eB144] (DyLight® 680)

Supplier: Rockland Immunochemicals
18-4417-32 18-4417-32
RL18-4417-32EA 296 USD InStock
CA18-4417-32 RL18-4417-32
Anti-IgG Rat Monoclonal Antibody [clone: eB144] (DyLight® 680)
Antibodies
Fluorescent Mouse TrueBlot® Antibody DyLight® 680 is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB144
Conjugation: DyLight® 680
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty