Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein A Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-077 200-103-077
RL200-103-077EA 339 USD InStock
CA200-103-077 RL200-103-077
Anti-Protein A Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Protein A Peroxidase Conjugated has been assayed against 1.0 µg of Protein A (Staphylococcus aureus) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:2000-1:20000; Western Blot: 1:500-1:2000; Immunochemistry: 1:250
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty