Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOXA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-093S 200-103-093S
RL200-103-093SEA 148 USD InStock
CA10036-750 RL200-103-093S
Anti-SOXA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Sarcosine Oxidase has been assayed against 1.0 µg of Sarcosine Oxidase (Microbial) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:1000 - 1:2500; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: SOXA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Microbial

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty