Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein A Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-077S 200-106-077S
RL200-106-077SEA 148 USD InStock
RL200-106-077S CA200-106-077S
Anti-Protein A Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Protein A Biotin Conjugated has been assayed against 1.0 µg of Protein A in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty