Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein A Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
200-109-077 200-109-077
RL200-109-077EA 339 USD InStock
CA200-109-077 RL200-109-077
Anti-Protein A Goat Polyclonal Antibody (Texas Red®)
Antibodies
Anti-Protein A Texa Red Conjugated Antibody is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty