Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LDHA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-1373S 200-1373S
RL200-1373SEA 148 USD InStock
RL200-1373S CA10036-858
Anti-LDHA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Lactate Dehydrogenase Antibody is suitable for western blotting, IP, Immunochemistryand for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:3000; Western Blot: 1:1000 - 1:6000; Immunochemistry: 1:500 - 1:2500; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Goat
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty