Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELA Mouse Monoclonal Antibody [clone: 27F9.G4]

Supplier: Rockland Immunochemicals
200-301-065 200-301-065S
RL200-301-065SEA 148 USD InStock
RL200-301-065S RL200-301-065
Anti-RELA Mouse Monoclonal Antibody [clone: 27F9.G4]
Antibodies
This Protein A purified mouse monoclonal antibody is directed against NFkB p65 (Rel A) and a 1:1000 dilution recognizes a 65 kD band by Western blot against HeLa whole cell lysate. Control peptide (100-4165p) is sold separately.

Recommended Dilutions: ELISA: 1:50000-1:100000; Immunofluorescence: 1:5000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:200-1:600;

Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 27F9.G4
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty