Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDKN1A Mouse Polyclonal Antibody [clone: WA-1]

Supplier: Rockland Immunochemicals
200-301-171 200-301-171
RL200-301-171EA 528 USD InStock
RL200-301-171 CA200-301-171
Anti-CDKN1A Mouse Polyclonal Antibody [clone: WA-1]
Antibodies
Primary Mouse Anti-p21/WAF1 Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN1A
Clonality: Polyclonal
Clone: WA-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty