Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GST Mouse Polyclonal Antibody [clone: 3D4]

Supplier: Rockland Immunochemicals
200-301-200 200-301-200
RL200-301-200EA 454 USD InStock
RL200-301-200 CA200-301-200
Anti-GST Mouse Polyclonal Antibody [clone: 3D4]
Antibodies
Primary Mouse Anti-Glutathione-S-Transferase (GST) Reacts with Pan

Type: Tag
Antigen: GST
Clonality: Monoclonal
Clone: 3D4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty