Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMGN1 Mouse Polyclonal Antibody [clone: IPO-38]

Supplier: Rockland Immunochemicals
200-301-278 200-301-278
RL200-301-278EA 528 USD InStock
RL200-301-278 CA200-301-278
Anti-HMGN1 Mouse Polyclonal Antibody [clone: IPO-38]
Antibodies
Anti-IPO-38 is suitable for ELISA,  immunoprecipitation, western blotting and immunochemistry (frozen and formalin/paraffin). The antibody is reported to recognize a nuclear antigen that is present in the cytoplasm and nuclei of proliferating cells.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty