Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Mouse Polyclonal Antibody [clone: 33D1.D2.G8]

Supplier: Rockland Immunochemicals
200-301-382 200-301-382S
RL200-301-382EA 362 USD InStock
RL200-301-382 RL200-301-382S
Anti-6xHis Mouse Polyclonal Antibody [clone: 33D1.D2.G8]
Antibodies
Anti-6X His is optimally suited for monitoring expression of His-tagged fusion Proteins. As such, anti-6X His/6X His can be used to identify fusion Proteins that contain the 6X His epitope.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:200;

Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone: 33D1.D2.G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty