Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]

Supplier: Rockland Immunochemicals
200-301-400S
RL200-301-400SEA 150 USD InStock
RL200-301-400S
Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]
Antibodies
Protein A Purified Mab anti-ATM has been tested by ELISA and western blotting against both the native and recombinant forms of the Protein. The antibody immunoprecipitates ATM from irradiated human and mouse cells.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Immunofluorescence: 1:100 - 1:500; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat,

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty