Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Monoclonal Antibody [clone: 14E5.16C8.25F6]

Supplier: Rockland Immunochemicals
200-301-401S
RL200-301-401SEA 148 USD InStock
RL200-301-401S
Anti-AKT1 Mouse Monoclonal Antibody [clone: 14E5.16C8.25F6]
Antibodies
Anti-AKT Antibody is suitable for ELISA, immunochemistry, immunoprecipitation and western blotting. Expect a band approximately 54 - 56 kDa in size corresponding to AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.16C8.25F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chimpanzee

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty