Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLH Mouse Monoclonal Antibody [clone: 12B4.G3.A8]

Supplier: Rockland Immunochemicals
200-301-466 200-301-466
RL200-301-466EA 333 USD InStock
RL200-301-466 CA200-301-466
Anti-KLH Mouse Monoclonal Antibody [clone: 12B4.G3.A8]
Antibodies
Anti-KLH can be used as a control in various assays or in ELISA assays to detect KLH or KLH conjugates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Tag
Antigen: KLH
Clonality: Monoclonal
Clone: 12B4.G3.A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty