Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TUBA1B Mouse Polyclonal Antibody [clone: 17H11.F10]

Supplier: Rockland Immunochemicals
200-301-880
RL200-301-880EA 415 USD InStock
RL200-301-880
Anti-TUBA1B Mouse Polyclonal Antibody [clone: 17H11.F10]
Antibodies
Type: Primary
Antigen: TUBA1B
Clonality: Polyclonal
Clone: 17H11.F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Chicken, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty