Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BUB1B Mouse Polyclonal Antibody [clone: 8G1]

Supplier: Rockland Immunochemicals
200-301-902 200-301-902
RL200-301-902EA 528 USD InStock
RL200-301-902 CA200-301-902
Anti-BUB1B Mouse Polyclonal Antibody [clone: 8G1]
Antibodies
This Protein A purified antibody has been tested for use in immunoprecipitation, immunofluorescence staining and western blot and is capable of detecting endogenous Protein. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: BUB1B
Clonality: Polyclonal
Clone: 8G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty