Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAD1L1 Mouse Polyclonal Antibody [clone: 9B10]

Supplier: Rockland Immunochemicals
200-301-903 200-301-903
RL200-301-903EA 528 USD InStock
RL200-301-903 CA200-301-903
Anti-MAD1L1 Mouse Polyclonal Antibody [clone: 9B10]
Antibodies
This Protein A purified antibody has been tested for use in flow cytometry, immunoprecipitation, immunofluorescence and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band at ~ 83 kDa.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MAD1L1
Clonality: Polyclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty