Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NEDD9 Mouse Polyclonal Antibody [clone: 2G9]

Supplier: Rockland Immunochemicals
200-301-904 200-301-904
RL200-301-904EA 528 USD InStock
RL200-301-904 CA200-301-904
Anti-NEDD9 Mouse Polyclonal Antibody [clone: 2G9]
Antibodies
This monoclonal antibody has been tested for use in western blotting, immunoprecipitation and immunofluorescence.  Specific conditions for reactivity should be optimized by the end user.  Expect bands approximately 115 and 105 in size.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:500; Western Blot: 1:5000; Immunoprecipitation: 1:1000;

Type: Primary
Antigen: NEDD9
Clonality: Polyclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty