Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NEDD9 Mouse Polyclonal Antibody [clone: 14A11]

Supplier: Rockland Immunochemicals
200-301-912 200-301-912
RL200-301-912EA 528 USD InStock
RL200-301-912 CA200-301-912
Anti-NEDD9 Mouse Polyclonal Antibody [clone: 14A11]
Antibodies
This monoclonal antibody has been tested for use in western blotting, immunoprecipitation and immunofluorescence.  This clone recognizes HEF1 under non-denaturing conditions.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:100; Western Blot: 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: NEDD9
Clonality: Polyclonal
Clone: 14A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty