Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDF15 Mouse Monoclonal Antibody [clone: 23G10.F8]

Supplier: Rockland Immunochemicals
200-301-B10S 200-301-B10
RL200-301-B10SEA 148 USD InStock
RL200-301-B10S RL200-301-B10
Anti-GDF15 Mouse Monoclonal Antibody [clone: 23G10.F8]
Antibodies
This Protein A purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.  For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF15 C-terminal specific antibodies.

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:1000;

Type: Primary
Antigen: NAG-1
Clonality: Monoclonal
Clone: 23G10.F8
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty