Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FLAG™ conjugated Proteins Mouse Polyclonal Antibody [clone: 29E4.G7]

Supplier: Rockland Immunochemicals
200-301-B13 200-301-B13 200-301-383 200-301-383
RL200-301-B13EA 587 USD InStock
CA200-301-383 RL200-301-B13 RL200-301-383 CA200-301-B13
Anti-FLAG™ conjugated Proteins Mouse Polyclonal Antibody [clone: 29E4.G7]
Antibodies
Primary Mouse Anti-FLAG™ conjugated proteins Reacts with Pan

Type: Primary
Antigen: FLAG™ conjugated Proteins
Clonality: Polyclonal
Clone: 29E4.G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty