Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]

Supplier: Rockland Immunochemicals
200-301-B45 200-301-B45S
RL200-301-B45SEA 148 USD InStock
RL200-301-B45S RL200-301-B45
Anti-LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Expect a band approximately 102 kDa in size corresponding to LGR4 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:500 - 1:3000; Immunochemistry: 5 µg/ml;

Type: Primary
Antigen: LGR4
Clonality: Monoclonal
Clone: 6G8.B3.G5.C3
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Chimpanzee, Macaque

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty