Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRPC6 Mouse Monoclonal Antibody [clone: 3F2.H10.F2]

Supplier: Rockland Immunochemicals
200-301-B59S 200-301-B59
RL200-301-B59SEA 148 USD InStock
RL200-301-B59S RL200-301-B59
Anti-TRPC6 Mouse Monoclonal Antibody [clone: 3F2.H10.F2]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Expect a band approximately 106 kDa in size corresponding to TRPC6 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 2.5 µg/ml;

Type: Primary
Antigen: TRPC6
Clonality: Monoclonal
Clone: 3F2.H10.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty