Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT2 Mouse Monoclonal Antibody [clone: 19G8.H2.H6]

Supplier: Rockland Immunochemicals
200-301-B65S 200-301-B65
RL200-301-B65SEA 148 USD InStock
RL200-301-B65S RL200-301-B65
Anti-STAT2 Mouse Monoclonal Antibody [clone: 19G8.H2.H6]
Antibodies
This Protein-A purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 113 kDa in size corresponding to STAT2 Protein by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:1000; Immunochemistry: 1:2.5 µg/ml;

Type: Primary
Antigen: STAT2
Clonality: Monoclonal
Clone: 19G8.H2.H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty