Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABCA4 Mouse Monoclonal Antibody [clone: 3F4]

Supplier: Rockland Immunochemicals
200-301-D05
RL200-301-D05EA 528 USD InStock
RL200-301-D05
Anti-ABCA4 Mouse Monoclonal Antibody [clone: 3F4]
Antibodies
Anti-ABCA4 antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions should be optimized by the end user. Expect a band of ~250kDa in size corresponding to ABCA4 Proteins in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: ABCA4
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Xenopus, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty