Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALCA Mouse Monoclonal Antibody [clone: 4901]

Supplier: Rockland Immunochemicals
200-301-D15
RL200-301-D15EA 528 USD InStock
RL200-301-D15
Anti-CALCA Mouse Monoclonal Antibody [clone: 4901]
Antibodies
Anti-Calcitonin Gene Related Peptide antibody is suitable for use in ELISA and ICC. This product is not recommended for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:5000; Immunofluorescence: 1:50-1:100; Immunochemistry: 1:50-1:100; Immunoprecipitation: 1:50-1:100

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 4901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty